2-Dimethylaminobenzylhydrazine dihydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

OR0200-250MG
APOSOR0200-250MGEA 112 EUR
APOSOR0200-250MG
2-Dimethylaminobenzylhydrazine dihydrochloride 95+%
2-Dimethylaminobenzylhydrazine dihydrochloride
Formel: C₉H₁₇Cl₂N₃
Molecular Weight: 238,16 g/mol
Schmelzpunkt: >165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245511
CAS-Nummer: 849021-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR