4-Hexylphenylhydrazin Hydrochlorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR0201-250MG
APOSOR0201-250MGEA
116
EUR
APOSOR0201-250MG
4-Hexylphenylhydrazin Hydrochlorid 95%
4-Hexylphenylhydrazin Hydrochlorid
Formel:
C₁₂H₂₁ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245514 CAS-Nummer: 126062-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...