4'-(1-Piperazinyl)acetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Piperazinoacetophenone, 1-(4-(Piperazin-1-yl)phenyl)ethanone
OR0219-25G
OR0219-5G
APOSOR0219-25GEA
318
EUR
APOSOR0219-25G
APOSOR0219-5G
4'-(1-Piperazinyl)acetophenon
4'-(1-Piperazinyl)acetophenon
Formel:
C₁₂H₁₆N₂O Schmelzpunkt: 107…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005962 CAS-Nummer: 51639-48-6 EINECS: 257-332-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...