1-Methyl-3-phenylpiperazin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: MEFPIP

OR0221-1G
APOSOR0221-1GEA 21.9 EUR
APOSOR0221-1G
1-Methyl-3-phenylpiperazin 97%
1-Methyl-3-phenylpiperazin
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
Schmelzpunkt: 60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411603
CAS-Nummer: 5271-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR