tert-Butyl (piperazin-1-yl)acetate dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR0238-5G
APOSOR0238-5GEA 0 EUR
APOSOR0238-5G
tert-Butyl (piperazin-1-yl)acetate dihydrochloride 97%
tert-Butyl (piperazin-1-yl)acetate dihydrochloride
Formel: C₁₀H₂₀N₂O₂.2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD04972649
CAS-Nummer: 827614-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR