4'-Chlor-2'-methylacetanilid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Chloro-2-methylphenyl)acetamide

OR0250-1G
APOSOR0250-1GEA 87 EUR
APOSOR0250-1G
4'-Chlor-2'-methylacetanilid 95%
4'-Chlor-2'-methylacetanilid
Formel: C₉H₁₀ClNO
Molecular Weight: 183,64 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00027399
CAS-Nummer: 5202-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR