4-Chlor-2-methylbenzonitril

Lieferant: Apollo Scientific
Achtung

OR0252-5G
APOSOR0252-5GEA 21.9 EUR
APOSOR0252-5G
4-Chlor-2-methylbenzonitril
4-Chlor-2-methylbenzonitril
Formel: C₈H₆ClN
Molecular Weight: 151,60 g/mol
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045603
CAS-Nummer: 50712-68-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR