2-Chlor-4-(methylsulfonyl)benzoesäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR0264-100G
OR0264-25G
OR0264-5G
APOSOR0264-100GEA
33.5
EUR
APOSOR0264-100G
APOSOR0264-25G
APOSOR0264-5G
2-Chlor-4-(methylsulfonyl)benzoesäure 97%
2-Chloro-4-(methylsulphonyl)benzoic acid
Formel:
C₈H₇ClO₄S Schmelzpunkt: 191…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216496 CAS-Nummer: 53250-83-2 EINECS: 406-520-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...