4-Chlor-3-nitrobenzolsulfonylchlorid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Chlor-3-nitrobenzolsulfonsäurechlorid, 4-Chlor-3-nitrobenzolsulfochlorid, 4-Chlor-3-nitrobenzol-1-sulfonylchlorid
OR0266-1G
APOSOR0266-1GEA
21.9
EUR
APOSOR0266-1G
4-Chlor-3-nitrobenzolsulfonylchlorid 98%
4-Chlor-3-nitrobenzolsulfonylchlorid
Formel:
C₆H₃Cl₂NO₄S Molecular Weight: 256,07 g/mol Schmelzpunkt: 59…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007440 CAS-Nummer: 97-08-5 EINECS: 202-558-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...