Benzo[b]furan-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR0275-1G OR0275-250MG
APOSOR0275-1GEA 97.3 EUR
APOSOR0275-1G APOSOR0275-250MG
Benzo[b]furan-2-carbohydrazide
Benzo[b]furan-2-carbohydrazide
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00456546
CAS-Nummer: 42974-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR