trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR0279-5G OR0279-1G OR0279-25G
APOSOR0279-5GEA 56.7 EUR
APOSOR0279-5G APOSOR0279-1G APOSOR0279-25G
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
Formel: C₁₃H₁₅ClO₂
Schmelzpunkt: 249…255 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797626
CAS-Nummer: 49708-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR