3-Chlor-6-phenylpyridazin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Phenyl-6-chloropyridazine

OR0288-1G
APOSOR0288-1GEA 40.6 EUR
APOSOR0288-1G
3-Chlor-6-phenylpyridazin 97%
3-Chloro-6-phenylpyridazine
Formel: C₁₀H₇ClN₂
Molecular Weight: 190,6300 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075253
CAS-Nummer: 20375-65-9
EINECS: 243-772-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR