3-Chloro-2,5-dihydrothiophene-1,1-dioxide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-3-thiolene 1,1-dioxide

OR0299-1G
APOSOR0299-1GEA 609 EUR
APOSOR0299-1G
3-Chloro-2,5-dihydrothiophene-1,1-dioxide
3-Chloro-2,5-dihydrothiophene-1,1-dioxide
Formel: C₄H₅ClO₂S
Molecular Weight: 152,6 g/mol
Schmelzpunkt: 98…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00154884
CAS-Nummer: 7311-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR