Cinnamaldoxime
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Cinnamaldehyde oxime
OR0305-1G
APOSOR0305-1GEA
21.9
EUR
APOSOR0305-1G
Cinnamaldoxime
Cinnamaldoxime
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Schmelzpunkt: 117…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019969 CAS-Nummer: 13372-81-1 EINECS: 236-449-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...