3-Oxabicyclo[3.2.0]heptane-2,4-dione 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Cyclobutane-1,2-dicarboxylic acid anhydride

OR0316-250MG
APOSOR0316-250MGEA 21.9 EUR
APOSOR0316-250MG
3-Oxabicyclo[3.2.0]heptane-2,4-dione 95+%
3-Oxabicyclo[3.2.0]heptane-2,4-dione
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
Schmelzpunkt: 73…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00799396
CAS-Nummer: 4462-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR