Cyclopentylchlorformiat 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Cyclopentyl carbonochloridate
OR0322-5G
APOSOR0322-5GEA
119
EUR
APOSOR0322-5G
Cyclopentylchlorformiat 97%
Cyclopentylchlorformiat
Formel:
C₆H₉ClO₂ Molecular Weight: 148.59 g/mol Siedepunkt: 65 °C/23mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00144029 CAS-Nummer: 50715-28-1 EINECS: 411-460-0 UN: 3277 ADR: 6.1+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...