L(+)-Cystein

Lieferant: Apollo Scientific
Achtung

OR0325-100G OR0325-1KG OR0325-250G
APOSOR0325-100GEA 0 EUR
APOSOR0325-100G APOSOR0325-1KG APOSOR0325-250G
L(+)-Cystein
L-Cystein
Formel: HSCH₂CH(NH₂)CO₂H
Lagertemperatur: Raumtemperatur
MDL: MFCD00064306
CAS-Nummer: 52-90-4
EINECS: 200-158-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR