2'-Deoxyuridin
Lieferant: Apollo Scientific
|
Achtung
|
OR0326-100G
OR0326-25G
OR0326-5G
APOSOR0326-100GEA
0
EUR
APOSOR0326-100G
APOSOR0326-25G
APOSOR0326-5G
2'-Deoxyuridin
2'-Deoxyuridin
Formel:
C₉H₁₂N₂O₅ Schmelzpunkt: 165…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006527 CAS-Nummer: 951-78-0 EINECS: 213-455-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...