3,4-Bis(acetamido)benzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Diacetamidobenzoic acid

OR0327-1G
APOSOR0327-1GEA 72.5 EUR
APOSOR0327-1G
3,4-Bis(acetamido)benzoesäure
3,4-Bis(acetamido)benzoesäure
Formel: C₁₁H₁₂N₂O₄
Schmelzpunkt: 226…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00238577
CAS-Nummer: 205873-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR