2,3-Dichlorbenzaldehydoxim

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dichlorobenzaldoxime

OR0337-1G
APOSOR0337-1GEA 42.2 EUR
APOSOR0337-1G
2,3-Dichlorbenzaldehydoxim
2,3-Dichlorbenzaldehydoxim
Formel: C₇H₅Cl₂NO
Molecular Weight: 190,03 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 4414-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR