3,5-Dichlorbenzhydrazid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichlorobenzoic hydrazide

OR0341-250MG
APOSOR0341-250MGEA 21.9 EUR
APOSOR0341-250MG
3,5-Dichlorbenzhydrazid
3,5-Dichlorbenzhydrazid
Formel: C₇H₆Cl₂N₂O
Molecular Weight: 205,04 g/mol
Schmelzpunkt: 205…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00833411
CAS-Nummer: 62899-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR