2',4'-Dichlor-6'-methylacetanilid

Lieferant: Apollo Scientific

OR0352-1G OR0352-5G OR0352-10G
APOSOR0352-1GEA 105 EUR
APOSOR0352-1G APOSOR0352-5G APOSOR0352-10G
2',4'-Dichlor-6'-methylacetanilid
2',4'-Dichlor-6'-methylacetanilid
Formel: C₉H₉Cl₂NO
Schmelzpunkt: 174…176 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 61655-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR