2,4-Dichlorophenoxyacetamidoxime

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,4-Dichlorophenoxy)-N'-hydroxyacetamidine

OR0358-250MG
APOSOR0358-250MGEA 101 EUR
APOSOR0358-250MG
2,4-Dichlorophenoxyacetamidoxime
2,4-Dichlorophenoxyacetamidoxime
Formel: C₈H₈Cl₂N₂O₂
Molecular Weight: 235,067 g/mol
Schmelzpunkt: 141…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02090614
CAS-Nummer: 79295-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR