(3,4-Dichlorphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,4-Dichlorophenylacetic acid
OR0359-500G
APOSOR0359-500GEA
111
EUR
APOSOR0359-500G
(3,4-Dichlorphenyl)essigsäure 98%
(3,4-Dichlorphenyl)essigsäure
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 314 Schmelzpunkt: 87…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004333 CAS-Nummer: 5807-30-7 EINECS: 227-368-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...