2,4-Dichlorphenylisocyanat

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,4-Dichlorophenylisocyantae

OR0363-5G
APOSOR0363-5GEA 62.5 EUR
APOSOR0363-5G
2,4-Dichlorphenylisocyanat
2,4-Dichlorphenylisocyanat

  • May form small amount of sediment.

Formel: C₇H₃Cl₂NO
Molecular Weight: 188.01 g/mol
Siedepunkt: 80 °C/1mmHg
Schmelzpunkt: 57…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002001
CAS-Nummer: 2612-57-9
EINECS: 220-040-4
UN: 2250
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR