2,3-Dihydrobenzo[b]furan-5-amidoxime 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-N'-hydroxy-1-benzofuran-5-carboximidamide, 2,3-Dihydro-N'-hydroxybenzo[b]furan-5-carboxamidine

OR0376-1G
APOSOR0376-1GEA 174 EUR
APOSOR0376-1G
2,3-Dihydrobenzo[b]furan-5-amidoxime 95+%
2,3-Dihydrobenzo[b]furan-5-amidoxime
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178,188 g/mol
Schmelzpunkt: 215…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04035721
CAS-Nummer: 306936-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR