2,5-Dimethoxybenzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
OR0402-250MG
APOSOR0402-250MGEA
21.9
EUR
APOSOR0402-250MG
2,5-Dimethoxybenzolsulfonamid
2,5-Dimethoxybenzolsulfonamid
Formel:
C₈H₁₁NO₄S Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00833415 CAS-Nummer: 19116-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...