1-Chlor-4-iod-2,5-dimethoxybenzol

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-2,5-dimethoxy-4-iodobenzene

OR0403-1G
APOSOR0403-1GEA 406 EUR
APOSOR0403-1G
1-Chlor-4-iod-2,5-dimethoxybenzol
1-Chlor-4-iod-2,5-dimethoxybenzol
Formel: C₈H₈ClIO₂
Molecular Weight: 298.51 g/mol
Schmelzpunkt: 115…116 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 90064-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR