3,5-Dimethyladamantane-1-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR0409-1G
OR0409-25G
OR0409-5G
APOSOR0409-1GEA
34.8
EUR
APOSOR0409-1G
APOSOR0409-25G
APOSOR0409-5G
3,5-Dimethyladamantane-1-carboxylic acid 95%
3,5-Dimethyladamantane-1-carboxylic acid
Formel:
C₁₃H₂₀O₂ Molecular Weight: 208.3 g/mol Schmelzpunkt: 89…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00188067 CAS-Nummer: 14670-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...