4-(Dimethylamino)phenylisothiocyanat
Lieferant: Apollo Scientific
|
Gefahr
|
OR0411-1G
OR0411-5G
OR0411-25G
APOSOR0411-1GEA
71.2
EUR
APOSOR0411-1G
APOSOR0411-5G
APOSOR0411-25G
4-(Dimethylamino)phenylisothiocyanat
4-(Dimethylamino)phenylisothiocyanat
Formel:
C₉H₁₀N₂S Schmelzpunkt: 64…66 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00041093 CAS-Nummer: 2131-64-8 EINECS: 218-360-4 UN: 2928 ADR: 6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...