1,3-Dimethyl-1H-pyrazol-5-carbonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
OR0419-1G
OR0419-250MG
APOSOR0419-1GEA
0
EUR
APOSOR0419-1G
APOSOR0419-250MG
1,3-Dimethyl-1H-pyrazol-5-carbonylchlorid
1,3-Dimethyl-1H-pyrazol-5-carbonylchlorid
Formel:
C₆H₇ClN₂O Siedepunkt: 100 °C/20mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00833258 CAS-Nummer: 55458-67-8 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...