1,3-Dimethyl-1H-pyrazole-5-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR0420-250MG
APOSOR0420-250MGEA
21.9
EUR
APOSOR0420-250MG
1,3-Dimethyl-1H-pyrazole-5-carboxamide
1,3-Dimethyl-1H-pyrazole-5-carboxamide
Formel:
C₆H₉N₃O Schmelzpunkt: 165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00833259 CAS-Nummer: 136678-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...