6-(Fmoc-amino)-1-hexanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 9-Fluorenylmethyl N-(6-hydroxyhexyl)carbamate, (9H-Fluoren-9-yl)methyl (6-hydroxyhexyl)carbamate, 6-Fmoc-Acp-ol

OR0485-1G
APOSOR0485-1GEA 0 EUR
APOSOR0485-1G
6-(Fmoc-amino)-1-hexanol
6-(Fmoc-amino)-1-hexanol
Formel: C₂₁H₂₅NO₃
Molecular Weight: 339,43 g/mol
Schmelzpunkt: 134…137 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00380196
CAS-Nummer: 127903-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR