N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine 95+%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-L-isoleucine, Fmoc-Ile-OH, FMOC-L-Isoleucine

OR0490-100G
APOSOR0490-100GEA 78.3 EUR
APOSOR0490-100G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine 95+%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,42 g/mol
Schmelzpunkt: 146…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037125
CAS-Nummer: 71989-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR