5-n-Hexylthio-1,3,4-thiadiazole-2-thiol
Lieferant: Apollo Scientific
OR0500-1G
APOSOR0500-1GEA
102
EUR
APOSOR0500-1G
5-n-Hexylthio-1,3,4-thiadiazole-2-thiol
5-n-Hexylthio-1,3,4-thiadiazole-2-thiol
Formel:
C₈H₁₄N₂S₃ Molecular Weight: 234,41 g/mol Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00186165 CAS-Nummer: 4858-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...