3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)-4-(3-carboxybenzyl)piperazine, tert-Butyl 4-(3-carboxybenzyl)piperazine-1-carboxylate

OR0508-250MG
APOSOR0508-250MGEA 190 EUR
APOSOR0508-250MG
3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
Formel: C₁₇H₂₄N₂O₄
Molecular Weight: 320,383 g/mol
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115219
CAS-Nummer: 500013-38-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR