2-(1,4-Diazepan-1-yl)ethanol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(2-Hydroxyethyl)homopiperazine

OR0518-1G
APOSOR0518-1GEA 125 EUR
APOSOR0518-1G
2-(1,4-Diazepan-1-yl)ethanol
2-(1,4-Diazepan-1-yl)ethanol
Formel: C₇H₁₆N₂O
Molecular Weight: 144,22 g/mol
Siedepunkt: 116…120 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01565842
CAS-Nummer: 53427-65-9
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR