3-(2-Hydroxyethyl)pyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR0520-1G
OR0520-5G
APOSOR0520-1GEA
58
EUR
APOSOR0520-1G
APOSOR0520-5G
3-(2-Hydroxyethyl)pyridin
3-(2-Hydroxyethyl)pyridine
Formel:
C₇H₉NO Siedepunkt: 176 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00831047 CAS-Nummer: 6293-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...