4-Hydroxy-7-methyl-3-phenyl-1H-quinolin-2-one

Lieferant: Apollo Scientific

OR0532-1G
APOSOR0532-1GEA 364 EUR
APOSOR0532-1G
4-Hydroxy-7-methyl-3-phenyl-1H-quinolin-2-one
4-Hydroxy-7-methyl-3-phenyl-1H-quinolin-2-one
Formel: C₁₆H₁₃NO₂
Molecular Weight: 251,28 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100772
CAS-Nummer: 83609-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR