Hypoxanthin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Purin-6-on, 6-Purinon, Purin-6(1H)-on, 6-Hydroxypurin

OR0545-100G
APOSOR0545-100GEA 21.9 EUR
APOSOR0545-100G
Hypoxanthin 98%
Hypoxanthin
Formel: C₅H₄N₄O
Molecular Weight: 136,11 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005725
CAS-Nummer: 68-94-0
EINECS: 200-697-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR