7-Iodo-9H-fluorene-4-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Iodo-9H-fluorene-5-carboxylic acid
OR0550-100MG
APOSOR0550-100MGEA
65.4
EUR
APOSOR0550-100MG
7-Iodo-9H-fluorene-4-carboxylic acid 95%
7-Iodo-9H-fluorene-4-carboxylic acid
Formel:
C₁₄H₉IO₂ Molecular Weight: 336,125 g/mol Schmelzpunkt: 265…267 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00829127 CAS-Nummer: 16218-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...