3-Isopropoxy-4-methoxybenzoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Carboxy-2-isopropoxyanisole, 4-Methoxy-3-[(prop-2-yl)oxy]benzoic acid
OR0557-250MG
APOSOR0557-250MGEA
42.2
EUR
APOSOR0557-250MG
3-Isopropoxy-4-methoxybenzoic acid
3-Isopropoxy-4-methoxybenzoic acid
Formel:
C₁₁H₁₄O₄ Schmelzpunkt: 135…137 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00831064 CAS-Nummer: 159783-29-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...