4-(Methylsulfonyl)phenol
Lieferant: Apollo Scientific
|
Achtung
|
OR0562-100G
OR0562-25G
OR0562-5G
APOSOR0562-100GEA
198
EUR
APOSOR0562-100G
APOSOR0562-25G
APOSOR0562-5G
4-(Methylsulfonyl)phenol
4-(Methylsulfonyl)phenol
Formel:
C₇H₈O₃S Schmelzpunkt: 82…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047806 CAS-Nummer: 14763-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...