4-Isopropyl-2-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Nitro-4-(prop-2-yl)aniline, 4-Amino-3-nitrocumene
OR0566-1G
APOSOR0566-1GEA
116
EUR
APOSOR0566-1G
4-Isopropyl-2-nitroaniline
4-Isopropyl-2-nitroaniline
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180.204 g/mol Siedepunkt: 141…143 °C (1 mm Hg) Schmelzpunkt: 35…37 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092920 CAS-Nummer: 63649-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...