4-Methoxy-3-propoxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR0589-1G
APOSOR0589-1GEA
184
EUR
APOSOR0589-1G
4-Methoxy-3-propoxybenzoic acid
4-Methoxy-3-propoxybenzoic acid
Formel:
C₁₁H₁₄O₄ Molecular Weight: 210.23 g/mol Schmelzpunkt: 150…153 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00831063 CAS-Nummer: 5922-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...