3-Methylbenzamidoxim
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Hydroxy-3-methylbenzimidamide
OR0604-1G
OR0604-5G
APOSOR0604-1GEA
45.1
EUR
APOSOR0604-1G
APOSOR0604-5G
3-Methylbenzamidoxim
3-Methylbenzamidoxim
Formel:
C₈H₁₀N₂O Schmelzpunkt: 87…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00833266 CAS-Nummer: 40067-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...