Methyl-2-chlorisonicotinat
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate
OR0610-100G
APOSOR0610-100GEA
72.5
EUR
APOSOR0610-100G
Methyl-2-chlorisonicotinat
Methyl-2-chlorisonicotinat
Formel:
C₇H₆ClNO₂ Molecular Weight: 171.5810 g/mol Schmelzpunkt: 32…36 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01765409 CAS-Nummer: 58481-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...