Methyl-4,5-dimethoxy-2-nitrobenzoat
Lieferant: Apollo Scientific
|
Achtung
|
OR0619-100G
OR0619-50G
OR0619-250G
APOSOR0619-100GEA
71.2
EUR
APOSOR0619-100G
APOSOR0619-50G
APOSOR0619-250G
Methyl-4,5-dimethoxy-2-nitrobenzoat
Methyl-4,5-dimethoxy-2-nitrobenzoat
Formel:
C₁₀H₁₁NO₆ Schmelzpunkt: 141…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007239 CAS-Nummer: 26791-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...