5-Methyl-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: AS057278, 5-Methylpyrazole-3-carboxylic acid

OR0650-1G
APOSOR0650-1GEA 0 EUR
APOSOR0650-1G
5-Methyl-1H-pyrazole-3-carboxylic acid
5-Methyl-1H-pyrazole-3-carboxylic acid
Formel: C5H6N2O2
Molecular Weight: 126.11 g/mol
Schmelzpunkt: 236…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00462235
CAS-Nummer: 402-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR