6-Methylchinoxalin

Lieferant: Apollo Scientific

OR0653-5G OR0653-100G OR0653-25G
APOSOR0653-5GEA 0 EUR
APOSOR0653-5G APOSOR0653-100G APOSOR0653-25G
6-Methylchinoxalin
6-Methylquinoxaline
Formel: C₉H₈N₂
Molecular Weight: 144.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041001
CAS-Nummer: 6344-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR